
nor-Binaltorphimine dihydrochloride

Synonym(s):
nor-BNI dihydrochloride
Empirical Formula (Hill Notation):
C40H43N3O6 · 2HCl
CAS Number:
Molecular Weight:
734.71
Products
Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Research Area
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing