Skip to Content
Merck
N-Caffeoyltyramine phyproof®Reference Substance

N-Caffeoyltyramine

N-Caffeoyltyramine phyproof®Reference Substance
Synonym(s):
N-trans-caffeoyltyramine, Typheramide
Empirical Formula (Hill Notation):
C17H17NO4
CAS Number:
Molecular Weight:
299.32

Products

Formula Weight
Manufacturer
Product Line
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing