Morin hydrate
Synonym(s):
2′,3,4′,5,7-Pentahydroxyflavone
Empirical Formula (Hill Notation):
C15H10O7 · xH2O
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Research Area
Application
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing