![Monoclonal Anti-polyHistidine–Agarose antibody produced in mouse clone HIS-1, purified immunoglobulin, PBS suspension, suitable for purification of HIS tagged recombinant proteins](/deepweb/assets/sigmaaldrich/product/images/251/042/71423750-7480-4440-870c-370559246c82/640/71423750-7480-4440-870c-370559246c82.jpg)
Monoclonal Anti-polyHistidine–Agarose antibody produced in mouse
![Monoclonal Anti-polyHistidine–Agarose antibody produced in mouse clone HIS-1, purified immunoglobulin, PBS suspension, suitable for purification of HIS tagged recombinant proteins](/deepweb/assets/sigmaaldrich/product/images/251/042/71423750-7480-4440-870c-370559246c82/640/71423750-7480-4440-870c-370559246c82.jpg)
Synonym(s):
Monoclonal Anti-polyHistidine, Monoclonal 6 His epitope tag, Monoclonal 6xHis-tag, Monoclonal HHHHHH epitope tag, Monoclonal Hexa His tag, Monoclonal His-tag, Monoclonal His6 tag, Monoclonal Histidine tagged, Monoclonal Poly-His-tag
Products
Product Category
Brand
Biological Source
Clonality
Isotype
Physical Form
Antibody Form
Conjugate
Quality Segment
Post Translational Modification
Showing 1-1 of 1