Skip to Content
Merck
vibrant-m

Monoclonal Anti-L-Selectin (CD62L) antibody produced in mouse

vibrant-m
Synonym(s):
Monoclonal Anti-CD62L, Anti-L-Selectin

Products

Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique