Skip to Content
Merck
Monoclonal Anti-β-Catenin antibody produced in mouse clone 6F9, ascites fluid

Monoclonal Anti-β-Catenin antibody produced in mouse

Monoclonal Anti-β-Catenin antibody produced in mouse clone 6F9, ascites fluid
Synonym(s):
Monoclonal Anti-β-Catenin

Products

Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Antibody Form
Research Area
Conjugate
Quality Segment
Application
Post Translational Modification
Species Reactivity
Technique