Skip to Content
Merck
Micro particles based on polystyrene size: 3 μm

Micro particles based on polystyrene

Micro particles based on polystyrene size: 3 μm
Synonym(s):
Latex beads from PS

Products

Product Category
Brand
Physical Form
Format
Quality Segment
Application
Particle Size (µm)
Reference Material Type
Special Grade
Showing 1-20 of 20
Compare
Product Number
Description
Pricing