Skip to Content
Merck
Methyl phenylacetate ≥98%, FCC, FG

Methyl phenylacetate

Methyl phenylacetate ≥98%, FCC, FG
Linear Formula:
C6H5CH2CO2CH3
CAS Number:
Molecular Weight:
150.17
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Product Line
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing