![Methyl 3-[[(2RS)-2-(ethylamino)propanoyl]amino]-4-methylthiophene-2- carboxylate hydrochloride certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/148/537/13d275e7-a494-4237-865f-2c06ce01223c/640/13d275e7-a494-4237-865f-2c06ce01223c.png)
Methyl 3-[[(2RS)-2-(ethylamino)propanoyl]amino]-4-methylthiophene-2- carboxylate hydrochloride
![Methyl 3-[[(2RS)-2-(ethylamino)propanoyl]amino]-4-methylthiophene-2- carboxylate hydrochloride certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/148/537/13d275e7-a494-4237-865f-2c06ce01223c/640/13d275e7-a494-4237-865f-2c06ce01223c.png)
Synonym(s):
Ethylarticaine hydrochloride
Empirical Formula (Hill Notation):
C12H18N2O3S · HCl
Molecular Weight:
306.81
Products
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Quality Segment
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1