![L-Lactic acid-1-13C API for Clinical Studies, ≥99 atom % 13C, ≥95% (CP)](/deepweb/assets/sigmaaldrich/product/structures/426/731/099d46ab-13c7-4064-938c-d459b0691c7e/640/099d46ab-13c7-4064-938c-d459b0691c7e.png)
L-Lactic acid-1-13C
![L-Lactic acid-1-13C API for Clinical Studies, ≥99 atom % 13C, ≥95% (CP)](/deepweb/assets/sigmaaldrich/product/structures/426/731/099d46ab-13c7-4064-938c-d459b0691c7e/640/099d46ab-13c7-4064-938c-d459b0691c7e.png)
Synonym(s):
(S)-2-Hydroxypropionic acid-1-13C, Sarcolactic acid-1-13C
Linear Formula:
CH3CH(OH)13CO2H
CAS Number:
Molecular Weight:
91.07
Products
Product Category
Brand
Sustainability
Feature
Formula Weight
Physical Form
Purity
Quality Segment
Isotope
Markush Class
Markush Group
Optical Isomer
Special Grade
Technique
Showing 1-2 of 2