Skip to Content
Merck
Linolenic acid analytical standard

Linolenic acid

Linolenic acid analytical standard
Synonym(s):
α-Lnn, cis,cis,cis-9,12,15-Octadecatrienoic acid
Linear Formula:
CH3(CH2CH=CH)3(CH2)7CO2H
CAS Number:
Molecular Weight:
278.43
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Format
Quality Segment
Functional Group
Application
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing