![L-Glutamine-1,2-13C2 endotoxin tested, 99 atom % 13C](/deepweb/assets/sigmaaldrich/product/structures/404/226/b5554bb0-8d4e-49ad-8ce9-9b7028ee29ad/640/b5554bb0-8d4e-49ad-8ce9-9b7028ee29ad.png)
L-Glutamine-1,2-13C2
![L-Glutamine-1,2-13C2 endotoxin tested, 99 atom % 13C](/deepweb/assets/sigmaaldrich/product/structures/404/226/b5554bb0-8d4e-49ad-8ce9-9b7028ee29ad/640/b5554bb0-8d4e-49ad-8ce9-9b7028ee29ad.png)
Linear Formula:
H2NCO(CH2)213CH(NH2)13CO2H
Molecular Weight:
148.13
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Isotope
Markush Class
Markush Group
Optical Isomer
Showing 1-2 of 2
Compare
Product Number
Description
Pricing