Levalbuterol hydrochloride
Synonym(s):
(R)-salbutamol hydrochloride, Levosalbutamol hydrochloride, Levalbuterol hydrochloride
Empirical Formula (Hill Notation):
C13H21NO3 · HCl
CAS Number:
Molecular Weight:
275.77
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade