![Iopamidol Related Compound A certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/images/292/453/e6f52c2f-5259-4d1c-ad0b-1920273baffe/640/e6f52c2f-5259-4d1c-ad0b-1920273baffe.jpg)
Iopamidol Related Compound A
![Iopamidol Related Compound A certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/images/292/453/e6f52c2f-5259-4d1c-ad0b-1920273baffe/640/e6f52c2f-5259-4d1c-ad0b-1920273baffe.jpg)
Synonym(s):
5-Amino-N,N′-bis[2-hydroxy-1-(hydroxymethyl)ethyl]-2,4,6-triiodobenzene-1,3-dicarboxamide, N,N′-Bis(1,3-dihydroxy-2-propyl)-5-amino-2,4,6-triiodoisophthalamide, N,N′-Bis[2-hydroxy-1-(hydroxymethyl)ethyl]-5-amino-2,4,6-triiodoisophthalamide
Empirical Formula (Hill Notation):
C14H18I3N3O6
CAS Number:
Molecular Weight:
705.02
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2