Skip to Content
KZ
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Hycon
®
Contact Slides
Products
Product Category
media (8)
accessories (1)
Brand
Millipore (8)
Physical Form
Chemical Composition
Quality Segment
Application
Microorganism Suitability
Package Size
Sterility
Sterilization Method
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
1.44023
Hycon
®
Contact Slides,
Tryptic Soy Agar
, Lecithin, Polysorbate 80, Histidine, for bacteria, for fungi, aseptically filled, suitable for surface monitoring
Expand
View Pricing
1.44021
Hycon
®
Contact Slides,
D/E (Dey/Engley) Neutralizing Agar
, Polysorbate 80, for bacteria, for fungi, sterile; γ-irradiated, double packed of 20 units, suitable for surface monitoring
Expand
View Pricing
1.44017
Hycon
®
Contact Slides,
Sabouraud Dextrose Agar, Polysorbate 80, Phosphatidycholine, for yeasts, for molds, sterile; aseptically filled, single packed of 20 units, suitable for surface monitoring
Expand
View Pricing
1.44022
Hycon
®
Contact Slides,
Tryptic Soy Agar, Lecithin, Histidine, Polysorbate 80, for bacteria, for yeasts, for molds, sterile; γ-irradiated, double packed of 20 or 100 units, suitable for surface monitoring
Expand
View Pricing
1.44018
Hycon
®
Contact Slides,
Rose-Bengal Chloramphenicol Agar, for yeasts, for molds, sterile; aseptically filled, single packed of 20 units, suitable for surface monitoring
Expand
View Pricing
1.44024
Hycon
®
Contact Slides,
Empty slides, stand alone, sterile, pack of 20 units, suitable for surface monitoring
Expand
View Pricing
1.44019
Hycon
®
Contact Slides,
MacConkey Agar, for enterobacteriaceae, sterile; aseptically filled, single packed of 20 units, suitable for surface monitoring
Expand
View Pricing
1.44016
Hycon
®
Contact Slides,
Sabouraud Dextrose Agar, Polysorbate 80, Phosphatidycholine, for yeasts, for molds, sterile; γ-irradiated, double packed of 20 units, suitable for surface monitoring
Expand
View Pricing