Ginsenoside Ro
Synonym(s):
(3β)-28-(β-D-Glucopyranosyloxy)-28-oxoolean-12-en-3-yl 2-O-β-D-glucopyranosyl-β-D-glucopyranosiduronic acid, Chikusetsusaponin V, Polysciasaponin P3, (3β)-28-(β-D-Glucopyranosyloxy)-28-oxoolean-12-en-3-yl 2-O-β-D-glucopyranosyl-β-D-glucopyranosiduronic acid, Chikusetsusaponin 5
Empirical Formula (Hill Notation):
C48H76O19
CAS Number:
Molecular Weight:
957.11
Products
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Quality Segment
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing