Skip to Content
Merck
Fmoc-OSu Novabiochem®

Fmoc-OSu

Fmoc-OSu Novabiochem®
Synonym(s):
Fmoc-OSu
Empirical Formula (Hill Notation):
C19H15NO5
CAS Number:
Molecular Weight:
337.33

Products

Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Functional Group
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing