Skip to Content
Merck
Flocoumafen PESTANAL®, analytical standard

Flocoumafen

Flocoumafen PESTANAL®, analytical standard
Empirical Formula (Hill Notation):
C33H25F3O4
CAS Number:
Molecular Weight:
542.54
EC Number:

Products

Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing