Ethyl valproate
Synonym(s):
Ethyl 2,2-dipropylacetate, Ethyl 2-propylpentanoate
Empirical Formula (Hill Notation):
C10H20O2
CAS Number:
Molecular Weight:
172.26
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing