![Ethyl 4-[(2-acetyloxybenzoyl)amino]benzoate certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/380/916/c5500cd3-78db-4efd-bc7a-8b24536b8f2b/640/c5500cd3-78db-4efd-bc7a-8b24536b8f2b.png)
Ethyl 4-[(2-acetyloxybenzoyl)amino]benzoate
![Ethyl 4-[(2-acetyloxybenzoyl)amino]benzoate certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/380/916/c5500cd3-78db-4efd-bc7a-8b24536b8f2b/640/c5500cd3-78db-4efd-bc7a-8b24536b8f2b.png)
Synonym(s):
4-[[2-(Acetyloxy)benzoyl]amino]benzoic acid ethyl ester
Empirical Formula (Hill Notation):
C18H17NO5
CAS Number:
Molecular Weight:
327.33
Products
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Quality Segment
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing