Skip to Content
Merck
(+)-Pinoresinol phyproof® Reference Substance

(+)-Pinoresinol

(+)-Pinoresinol phyproof® Reference Substance
Synonym(s):
Pinoresinol, (+)-Pinoresinol, 4,4′-((1S,3aR,4S,6aR)-Hexahydrofuro[3,4-c]furan-1,4-diyl)bis(2-methoxyphenol), 4,4′-[(1S,3aR,4S,6aR)-Tetrahydro-1H,3H-furo[3,4-c]furan-1,4-diyl]bis(2-methoxyphenol)
Empirical Formula (Hill Notation):
C20H22O6
CAS Number:
Molecular Weight:
358.39

Products

Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing