Skip to Content
Merck
Doxofylline ≥98% (HPLC)

Doxofylline

Doxofylline ≥98% (HPLC)
Synonym(s):
7-(1,3-Dioxolan-2-ylmethyl)theophylline
Empirical Formula (Hill Notation):
C11H14N4O4
CAS Number:
Molecular Weight:
266.25

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing