Dofetilide
Synonym(s):
N-[4-[2-[Methyl[2-[4-[(methylsulfonyl)amino]phenoxy]ethyl]amino]ethyl]phenyl]methanesulfonamide, UK-68798, Dofetilide
Empirical Formula (Hill Notation):
C19H27N3O5S2
CAS Number:
Molecular Weight:
441.56
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3