Diquat dibromide monohydrate
Synonym(s):
Diquat
Empirical Formula (Hill Notation):
C12H12Br2N2 · H2O
CAS Number:
Molecular Weight:
362.06
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing