Skip to Content
Merck
Cycloheximide solution 0.1%, suitable for microbiology

Cycloheximide solution

Cycloheximide solution 0.1%, suitable for microbiology
Synonym(s):
CHX
Empirical Formula (Hill Notation):
C15H23NO4
CAS Number:
Molecular Weight:
281.35

Products

Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Physical Form
Activity Spectrum
Quality Segment
Application
Microorganism Suitability
Mode of Action
Sterility