Skip to Content
Merck
Cimifugin phyproof® Reference Substance

Cimifugin

Cimifugin phyproof® Reference Substance
Synonym(s):
(S)-7-(Hydroxymethyl)-2-(2-hydroxypropan-2-yl)-4-methoxy-2,3-dihydro-5H-furo[3,2-g]chromen-5-one, 2,3-Dihydro-4-methoxy-7-(hydroxymethyl)-2-(1-hydroxy-1-methylethyl)-5H-furo[3,2-g][1]benzopyran-5-one, Cimitin
Empirical Formula (Hill Notation):
C16H18O6
CAS Number:
Molecular Weight:
306.31

Products

Formula Weight
Manufacturer
Product Line
Purity
Agency
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2