Skip to Content
Merck
Chloramphenicol for peak identification European Pharmacopoeia (EP) Reference Standard

Chloramphenicol for peak identification

Chloramphenicol for peak identification European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Chloramphenicol, D-(−)-threo-2,2-Dichloro-N-[β-hydroxy-α-(hydroxymethyl)-β-(4-nitrophenyl)ethyl]acetamide, D-(−)-threo-2-Dichloroacetamido-1-(4-nitrophenyl)-1,3-propanediol, D-threo-2,2-Dichloro-N-[β-hydroxy-α-(hydroxymethyl)-4-nitrophenethyl]acetamide, Chloromycetin
Linear Formula:
Cl2CHCONHCH(CH2OH)CH(OH)C6H4NO2
CAS Number:
Molecular Weight:
323.13
EC Number:

Products

Formula Weight
Melting Point (°C)
API Family
Reference Material Type
Showing 1-1 of 1