Skip to Content
Merck
Calcium chloride

Calcium chloride

Calcium chloride
Synonym(s):
Calcium chloride dihydrate
Empirical Formula (Hill Notation):
CaCl2 · 2H2O
CAS Number:
Molecular Weight:
147.01
EC Number:

Products

Product Category
Brand
Feature
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing