![Boc-Sar-OH Novabiochem®](/deepweb/assets/sigmaaldrich/product/images/539/965/f8de361a-3019-4318-a7cb-903be76ed063/640/f8de361a-3019-4318-a7cb-903be76ed063.jpg)
Boc-Sar-OH
![Boc-Sar-OH Novabiochem®](/deepweb/assets/sigmaaldrich/product/images/539/965/f8de361a-3019-4318-a7cb-903be76ed063/640/f8de361a-3019-4318-a7cb-903be76ed063.jpg)
Synonym(s):
Boc-Sar-OH
Empirical Formula (Hill Notation):
C8H15NO4
CAS Number:
Molecular Weight:
189.21
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Application
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing