Skip to Content
Merck
Bis(trimethylsilyl)acetylene 99%

Bis(trimethylsilyl)acetylene

Bis(trimethylsilyl)acetylene 99%
Synonym(s):
BTMSA
Linear Formula:
(CH3)3SiC≡CSi(CH3)3
CAS Number:
Molecular Weight:
170.40
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing