Skip to Content
Merck
Benzonase® Nuclease, ultrapure ≥250 units/μL, ≥99% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous glycerol solution, ultrapure grade

Benzonase® Nuclease, ultrapure

Benzonase® Nuclease, ultrapure ≥250 units/μL, ≥99% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous glycerol solution, ultrapure grade
Synonym(s):
Endonuclease from Serratia marcescens
CAS Number:

Products

Product Category
Brand
Biological Source
Physical Form
Purity
Recombinant Host
Quality Segment
Application