Skip to Content
Merck
Baclofen solution 500 μg/mL (MeOH:H2O (10:90)(v/v)), certified reference material, ampule of 1 mL, Cerilliant®

Baclofen solution

Baclofen solution 500 μg/mL (MeOH:H2O (10:90)(v/v)), certified reference material, ampule of 1 mL, Cerilliant®
Synonym(s):
(±)-Baclofen, (±)-β-(Aminomethyl)-4-chlorobenzenepropanoic acid, Lioresal
Empirical Formula (Hill Notation):
C10H12ClNO2
CAS Number:
Molecular Weight:
213.66

Products

Brand
Feature
Formula Weight
Manufacturer
Format
Quality Segment
Package Size
Special Grade