Ascentis® Express 160 Å Phenyl-Hexyl (2.7 μm) HPLC Columns
Synonym(s):
Ascentis® Express 160 Å Phenyl-Hexyl, 2.7 μm HPLC Column, Ascentis® Express Phenyl-Hexyl, 2.7 μm HPLC Column
Products
Product Category
Brand
pH Value
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Quality Segment
Column Material
Package Size
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-16 of 16
Compare
Product Number
Description
Pricing