Asarylaldehyde
Synonym(s):
2,4,5-Trimethoxybenzaldehyde, Asaronaldehyde
Linear Formula:
(CH3O)3C6H2CHO
CAS Number:
Molecular Weight:
196.20
Products
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing