Skip to Content
Merck
α-Phellandrene natural, ≥85%, FG

α-Phellandrene

α-Phellandrene natural, ≥85%, FG
Synonym(s):
2-Methyl-5-(1-methylethyl)-1,3-cyclohexadiene, α-Phellandrene
Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23

Products

Product Category
Brand
Sustainability
Biological Source
Feature
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing