Anti-γ-Tubulin-Cy3™ antibody produced in rabbit
Synonym(s):
Anti-γ-Tubulin
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing