![Anti-GroEL antibody produced in rabbit IgG fraction of antiserum, buffered aqueous solution](/deepweb/assets/sigmaaldrich/product/images/311/548/f9446bd0-01a5-43f7-ab5d-04d402d1bddd/640/f9446bd0-01a5-43f7-ab5d-04d402d1bddd.jpg)
Anti-GroEL antibody produced in rabbit
![Anti-GroEL antibody produced in rabbit IgG fraction of antiserum, buffered aqueous solution](/deepweb/assets/sigmaaldrich/product/images/311/548/f9446bd0-01a5-43f7-ab5d-04d402d1bddd/640/f9446bd0-01a5-43f7-ab5d-04d402d1bddd.jpg)
Synonym(s):
Anti-GroEL
Products
Product Category
Brand
Biological Source
Clonality
Physical Form
Antibody Form
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing