Skip to Content
Merck
Anti-EIF4ENIF1 antibody produced in rabbit Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Anti-EIF4ENIF1 antibody produced in rabbit

Anti-EIF4ENIF1 antibody produced in rabbit Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Synonym(s):
Anti-4E-T antibody produced in rabbit, Anti-Eukaryotic translation initiation factor 4E nuclear import factor 1 antibody produced in rabbit, Anti-Eukaryotic translation initiation factor 4E transporter antibody produced in rabbit, Anti-eIF4E transporter antibody produced in rabbit, Anti-4E-T, Anti-Clast4, Anti-Eukaryotic translation initiation factor 4E nuclear import factor 1, Anti-Eukaryotic translation initiation factor 4E transporter, Anti-eIF4E-T

Products

Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique