
Anti-DGCR14 antibody produced in rabbit

Synonym(s):
Anti-DiGeorge syndrome critical region 14 antibody produced in rabbit, Anti-ES2 protein antibody produced in rabbit, Anti-Protein DGCR14 antibody produced in rabbit
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing