Anti-ARHGAP1 antibody produced in rabbit
Synonym(s):
Anti-ρ GTPase-activating protein 1 antibody produced in rabbit, Anti-ρ-Related small GTPase protein activator antibody produced in rabbit, Anti-CDC42 GTPase-activating protein antibody produced in rabbit, Anti-GTPase-activating protein rhoOGAP antibody produced in rabbit, Anti-p50-RhoGAP antibody produced in rabbit
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing