Skip to Content
Merck
vibrant-m

Amino methacrylate copolymer

vibrant-m
Synonym(s):
2-Methyl-2-propenoic acid butyl ester polymer with 2-(dimethylamino)ethyl 2-methyl-2-propenoate and methyl 2-methyl-2-propenoate
CAS Number:

Products

Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1