Skip to Content
Merck
Aloperine ≥98% (HPLC)

Aloperine

Aloperine ≥98% (HPLC)
Synonym(s):
(6R,6aR,13R,13aS)-1,3,4,6,6a,7,8,9,10,12,13,13a-Dodecahydro-6,13-methano-2H-dipyrido[1,2-a:3′,2′-e]azocine, Ormosanine
Empirical Formula (Hill Notation):
C15H24N2
CAS Number:
Molecular Weight:
232.36

Products

Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing