Skip to Content
Merck
7-Amino-4-methylcoumarin 99%

7-Amino-4-methylcoumarin

7-Amino-4-methylcoumarin 99%
Synonym(s):
Coumarin 120, 7-Amino-4-methylcoumarin
Empirical Formula (Hill Notation):
C10H9NO2
CAS Number:
Molecular Weight:
175.18

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group