![6-(γ,γ-Dimethylallylamino)purine suitable for plant cell culture, BioReagent, ≥90%](/deepweb/assets/sigmaaldrich/product/structures/131/789/3aa38636-edd2-4fce-9962-124d81f1d120/640/3aa38636-edd2-4fce-9962-124d81f1d120.png)
6-(γ,γ-Dimethylallylamino)purine
![6-(γ,γ-Dimethylallylamino)purine suitable for plant cell culture, BioReagent, ≥90%](/deepweb/assets/sigmaaldrich/product/structures/131/789/3aa38636-edd2-4fce-9962-124d81f1d120/640/3aa38636-edd2-4fce-9962-124d81f1d120.png)
Synonym(s):
2iP, N6-(2-Isopentenyl)adenine
Empirical Formula (Hill Notation):
C10H13N5
CAS Number:
Molecular Weight:
203.24
Products
Brand
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Application
Special Grade
Sterilization Method
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing