![4-Phenyl-1-cyclohexene analytical standard](/deepweb/assets/sigmaaldrich/product/structures/415/245/41cb9a8b-1e08-4b4a-b435-9defc86072aa/640/41cb9a8b-1e08-4b4a-b435-9defc86072aa.png)
4-Phenyl-1-cyclohexene
![4-Phenyl-1-cyclohexene analytical standard](/deepweb/assets/sigmaaldrich/product/structures/415/245/41cb9a8b-1e08-4b4a-b435-9defc86072aa/640/41cb9a8b-1e08-4b4a-b435-9defc86072aa.png)
Synonym(s):
3-Cyclohexen-1-ylbenzene, 4-Phenylcyclohexene
Empirical Formula (Hill Notation):
C12H14
CAS Number:
Molecular Weight:
158.24
Products
Brand
Formula Weight
Purity
Format
Quality Segment
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing