Skip to Content
Merck
3-Carene analytical standard

3-Carene

3-Carene analytical standard
Synonym(s):
δ3-Carene, 3,7,7-Trimethylbicyclo[4.1.0]hept-3-ene
Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing