Skip to Content
Merck
3,5-Dimethylisoxazol-4-yl-4-boronic acid ≥95%

3,5-Dimethylisoxazol-4-yl-4-boronic acid

3,5-Dimethylisoxazol-4-yl-4-boronic acid ≥95%
Empirical Formula (Hill Notation):
C5H8BNO3
CAS Number:
Molecular Weight:
140.93

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing