Skip to Content
Merck
2-Hydroxyethylflurazepam solution 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

2-Hydroxyethylflurazepam solution

2-Hydroxyethylflurazepam solution 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Empirical Formula (Hill Notation):
C17H14ClFN2O2
CAS Number:
Molecular Weight:
332.76
EC Number:

Products

Brand
Formula Weight
Manufacturer
Physical Form
Format
Application
Package Size
Reference Material Type
Special Grade
Technique