Skip to Content
Merck
vibrant-m

2-{5-[(Pyrrolidin-1-ylsulfonyl)methyl]-1H-indol-3-yl}ethanamine hemifumarate salt

vibrant-m
Empirical Formula (Hill Notation):
C15H21N3O2S · 0.5C4H4O4
Molecular Weight:
365.45

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade