![10-Bromo-5H-dibenz[b,f]azepine-5-carboxamide certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/132/139/67106d7f-f898-4ea3-a56a-114aaf2789f3/640/67106d7f-f898-4ea3-a56a-114aaf2789f3.png)
10-Bromo-5H-dibenz[b,f]azepine-5-carboxamide
![10-Bromo-5H-dibenz[b,f]azepine-5-carboxamide certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/132/139/67106d7f-f898-4ea3-a56a-114aaf2789f3/640/67106d7f-f898-4ea3-a56a-114aaf2789f3.png)
Synonym(s):
10-Bromocarbamazepine
Empirical Formula (Hill Notation):
C15H11BrN2O
CAS Number:
Molecular Weight:
315.16
Products
Brand
Formula Weight
Manufacturer
Quality Segment
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing